Search
Search

CFAP70| Gene Symbol: CFAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503856.4 | XP_001503906.1 | 2568 | 99 | ||
| XM_023645010.1 | XP_023500778.1 | 2587 | 99 | |||
| XM_023645015.1 | XP_023500783.1 | 2790 | 99 | |||
| XM_023645023.1 | XP_023500791.1 | 2687 | 99 | |||
| XM_023645035.1 | XP_023500803.1 | 2587 | 99 | |||
| XM_023645041.1 | XP_023500809.1 | 2461 | 99 | |||
| XM_023645050.1 | XP_023500818.1 | 2452 | 99 | |||
| XM_023645053.1 | XP_023500821.1 | 2392 | 99 | |||
| XM_023645058.1 | XP_023500826.1 | 2248 | 99 | |||
| XM_023645060.1 | XP_023500828.1 | 2407 | 99 | |||
Set Membership: |
|