Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ec07041676_m1
See other CAMK2G GE Assays ›
Gene Symbol:
CAMK2G
Gene Name:
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases:
-
Chromosome Location:
Chr.1: 61917345 - 61972629 on Build EquCab3.0
Species:
Horse
Species Specific ID (Flybase ID):
-
Assay ID
Ec07041676_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CAMK2G
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_001503918.6
XP_001503968.2
344
88
XM_003363484.4
XP_003363532.1
343
88
XM_003363486.4
XP_003363534.1
344
88
XM_003363487.4
XP_003363535.1
344
88
XM_005602491.2
XP_005602548.1
346
88
XM_005602494.3
XP_005602551.1
344
88
XM_005602496.3
XP_005602553.1
342
88
XM_014733241.2
XP_014588727.1
346
88
XM_023645387.1
XP_023501155.1
372
88
XM_023645392.1
XP_023501160.1
371
88
XM_023645400.1
XP_023501168.1
369
88
XM_023645416.1
XP_023501184.1
371
88
XM_023645421.1
XP_023501189.1
323
88
XM_023645428.1
XP_023501196.1
367
88
XM_023645436.1
XP_023501204.1
370
88
XM_023645441.1
XP_023501209.1
368
88
XM_023645447.1
XP_023501215.1
345
88
XM_023645453.1
XP_023501221.1
368
88
XM_023645471.1
XP_023501239.1
365
88
XM_023645477.1
XP_023501245.1
367
88
XM_023645483.1
XP_023501251.1
377
88
XM_023645489.1
XP_023501257.1
377
88
XM_023645500.1
XP_023501268.1
365
88
XM_023645503.1
XP_023501271.1
344
88
XM_023645509.1
XP_023501277.1
360
88
XM_023645517.1
XP_023501285.1
343
88
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Equus caballus
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline