Search
Search

KAT6B| Gene Symbol: KAT6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023645567.1 | XP_023501335.1 | 2841 | 99 | ||
| XM_023645576.1 | XP_023501344.1 | 2835 | 99 | |||
| XM_023645577.1 | XP_023501345.1 | 3132 | 99 | |||
| XM_023645584.1 | XP_023501352.1 | 2292 | 99 | |||
| XM_023645590.1 | XP_023501358.1 | 1615 | 99 | |||
| XM_023645594.1 | XP_023501362.1 | 1965 | 99 | |||
Set Membership: |
|