Search
Search

RASGEF1A| Gene Symbol: RASGEF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490265.5 | XP_001490315.2 | 798 | 59 | ||
| XM_023646707.1 | XP_023502475.1 | 800 | 59 | |||
| XM_023646712.1 | XP_023502480.1 | 782 | 59 | |||
| XM_023646716.1 | XP_023502484.1 | 12722 | 59 | |||
| XM_023646725.1 | XP_023502493.1 | 800 | 59 | |||
Set Membership: |
|