Search
Search

WDFY4| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014733715.2 | XP_014589201.2 | 4538 | 60 | ||
| XM_023648246.1 | XP_023504014.1 | 4573 | 60 | |||
| XM_023648250.1 | XP_023504018.1 | 4573 | 60 | |||