Search
Search

PRPF3| Gene Symbol: PRPF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003365037.4 | XP_003365085.1 | 905 | 85 | ||
| XM_005610169.3 | XP_005610226.1 | 938 | 85 | |||
| XM_005610170.3 | XP_005610227.1 | 901 | 85 | |||
| XM_005610172.3 | XP_005610229.1 | 871 | 85 | |||
| XM_023641251.1 | XP_023497019.1 | 730 | 85 | |||
| XM_023641252.1 | XP_023497020.1 | 969 | 85 | |||
Set Membership: |
|