Search
Search

PDZK1| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001496160.6 | XP_001496210.3 | 1000 | 76 | ||
| XM_014739893.2 | XP_014595379.1 | 703 | 76 | |||
| XM_023641311.1 | XP_023497079.1 | 700 | 76 | |||
| XM_023641312.1 | XP_023497080.1 | 709 | 76 | |||
Set Membership: |
|