Search
Search

AP4B1| Gene Symbol: AP4B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610305.3 | XP_005610362.1 | 2019 | 67 | ||
| XM_005610306.3 | XP_005610363.1 | 2006 | 67 | |||
| GenBank mRNA | JL627158.1 | - | 6 - 7 | 1235 | 67 | |
Set Membership: |
|