Search
Search

PSMA4| Gene Symbol: PSMA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001489071.6 | XP_001489121.3 | 369 | 127 | ||
| XM_023652692.1 | XP_023508460.1 | 250 | 127 | |||
| GenBank mRNA | JL616146.2 | - | 4 - 5 | 347 | 127 | |
Set Membership: |
|