Search
Search

LMAN1L| Gene Symbol: LMAN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023653550.1 | XP_023509318.1 | 2232 | 64 | ||
| XM_023653555.1 | XP_023509323.1 | 2232 | 64 | |||
| XM_023653561.1 | XP_023509329.1 | 2232 | 64 | |||
| XM_023653569.1 | XP_023509337.1 | 2232 | 64 | |||
| XM_023653579.1 | XP_023509347.1 | 6803 | 64 | |||
Set Membership: |
|