Search
Search
| Gene Symbol: NEO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654054.1 | XP_023509822.1 | 1264 | 97 | ||
| XM_023654058.1 | XP_023509826.1 | 1263 | 97 | |||
| XM_023654061.1 | XP_023509829.1 | 1263 | 97 | |||
| XM_023654068.1 | XP_023509836.1 | 1264 | 97 | |||
| XM_023654073.1 | XP_023509841.1 | 1262 | 97 | |||
| XM_023654076.1 | XP_023509844.1 | 1262 | 97 | |||
| XM_023654078.1 | XP_023509846.1 | 1265 | 97 | |||
| XM_023654080.1 | XP_023509848.1 | 1265 | 97 | |||
| XM_023654083.1 | XP_023509851.1 | 1266 | 97 | |||
| XM_023654089.1 | XP_023509857.1 | 1266 | 97 | |||
| XM_023654093.1 | XP_023509861.1 | 1268 | 97 | |||