Search
Search

TMOD2| Gene Symbol: TMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023617377.1 | XP_023473145.1 | 701 | 92 | ||
| XM_023617385.1 | XP_023473153.1 | 704 | 92 | |||
| XM_023617388.1 | XP_023473156.1 | 585 | 92 | |||
| XM_023617392.1 | XP_023473160.1 | 701 | 92 | |||
| XM_023617396.1 | XP_023473164.1 | 438 | 92 | |||
| XM_023617401.1 | XP_023473169.1 | 578 | 92 | |||
Set Membership: |
|