Search
Search

CATSPER2| Gene Symbol: CATSPER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918217.4 | XP_001918252.1 | 1251 | 104 | ||
| XM_005603078.3 | XP_005603135.1 | 1138 | 104 | |||
| XM_005603079.3 | XP_005603136.1 | 1057 | 104 | |||
| XM_014733758.2 | XP_014589244.1 | 943 | 104 | |||
Set Membership: |
|