Search
Search

CAPN3| Gene Symbol: CAPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503274.4 | XP_001503324.1 | 2499 | 93 | ||
| XM_001503281.4 | XP_001503331.1 | 2229 | 93 | |||
| XM_014733631.2 | XP_014589117.1 | 2388 | 93 | |||
| XM_014733632.2 | XP_014589118.1 | 2369 | 93 | |||
| XM_014733633.2 | XP_014589119.1 | 2359 | 93 | |||
| XM_023619326.1 | XP_023475094.1 | 1653 | 93 | |||
Set Membership: |
|