Search
Search

CAPN3| Gene Symbol: CAPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503274.4 | XP_001503324.1 | 1429 | 88 | ||
| XM_001503281.4 | XP_001503331.1 | 1291 | 88 | |||
| XM_014733631.2 | XP_014589117.1 | 1432 | 88 | |||
| XM_014733632.2 | XP_014589118.1 | 1431 | 88 | |||
| XM_014733633.2 | XP_014589119.1 | 1289 | 88 | |||
| XM_023619326.1 | XP_023475094.1 | 583 | 88 | |||
Set Membership: |
|