Search
Search

KNSTRN| Gene Symbol: KNSTRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503549.5 | XP_001503599.2 | 1156 | 67 | ||
| XM_023620015.1 | XP_023475783.1 | 1195 | 67 | |||
| XM_023620018.1 | XP_023475786.1 | 1205 | 67 | |||
Set Membership: |
|