Search
Search

PARP2| Gene Symbol: PARP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603175.3 | XP_005603232.1 | 1078 | 100 | ||
| XM_014734015.2 | XP_014589501.1 | 1154 | 100 | |||
| XM_014734016.2 | XP_014589502.1 | 1070 | 100 | |||
Set Membership: |
|