Search
Search

SLC39A2| Gene Symbol: SLC39A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014733822.2 | XP_014589308.1 | 1276 | 84 | ||
| XM_014733827.2 | XP_014589313.1 | 1276 | 84 | |||
| XM_023621158.1 | XP_023476926.1 | 1276 | 84 | |||
| XM_023621167.1 | XP_023476935.1 | 1276 | 84 | |||
Set Membership: |
|