Search
Search

GEMIN2| Gene Symbol: GEMIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492973.6 | XP_001493023.1 | 582 | 122 | ||
| XM_005603448.3 | XP_005603505.1 | 583 | 122 | |||
| XM_014734189.2 | XP_014589675.1 | 583 | 122 | |||
| XM_023624326.1 | XP_023480094.1 | 575 | 122 | |||
Set Membership: |
|