Search
Search

FAAP24| Gene Symbol: FAAP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490021.4 | XP_001490071.1 | 769 | 60 | ||
| XM_005596125.3 | XP_005596182.1 | 697 | 60 | |||
| XM_014733054.2 | XP_014588540.1 | 696 | 60 | |||
| XM_023649492.1 | XP_023505260.1 | 699 | 60 | |||
| XM_023649493.1 | XP_023505261.1 | 252 | 60 | |||
Set Membership: |
|