Search
Search

GRAMD1A| Gene Symbol: GRAMD1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649527.1 | XP_023505295.1 | 1295 | 64 | ||
| XM_023649528.1 | XP_023505296.1 | 1206 | 64 | |||
| XM_023649529.1 | XP_023505297.1 | 1294 | 64 | |||
| XM_023649530.1 | XP_023505298.1 | 902 | 64 | |||
| XM_023649531.1 | XP_023505299.1 | 1045 | 64 | |||
| XM_023649532.1 | XP_023505300.1 | 1045 | 64 | |||
Set Membership: |
|