Search
Search

C10H19orf54| Gene Symbol: C10H19orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498105.5 | XP_001498155.2 | 1543 | 90 | ||
| XM_014736502.2 | XP_014591988.1 | 1543 | 90 | |||
| XM_023649778.1 | XP_023505546.1 | 755 | 90 | |||