Search
Search

TRAPPC6A| Gene Symbol: TRAPPC6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005596342.3 | XP_005596399.2 | 516 | 64 | ||
| XM_005596343.3 | XP_005596400.1 | 397 | 64 | |||
| XM_023649891.1 | XP_023505659.1 | 737 | 64 | |||
| XM_023649892.1 | XP_023505660.1 | 676 | 64 | |||
Set Membership: |
|