Search
Search
| Gene Symbol: VASP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023649915.1 | XP_023505683.1 | 449 | 67 | ||
| XM_023649916.1 | XP_023505684.1 | 451 | 67 | |||
| XM_023649917.1 | XP_023505685.1 | 450 | 67 | |||
| XM_023649918.1 | XP_023505686.1 | 452 | 67 | |||
Set Membership: |
|