Search
Search

AKT1S1| Gene Symbol: AKT1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493191.5 | XP_001493241.2 | 603 | 118 | ||
| XM_005596413.3 | XP_005596470.2 | 1019 | 118 | |||
| XM_023650191.1 | XP_023505959.1 | 639 | 118 | |||
Set Membership: |
|