Search
Search
| Gene Symbol: MZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651532.1 | XP_023507300.1 | 1639 | 79 | ||
| XM_023651533.1 | XP_023507301.1 | 1558 | 79 | |||
| XM_023651534.1 | XP_023507302.1 | 2080 | 79 | |||
| XM_023651535.1 | XP_023507303.1 | 2434 | 79 | |||
| XM_023651536.1 | XP_023507304.1 | 1731 | 79 | |||
| XM_023651537.1 | XP_023507305.1 | 1369 | 79 | |||
| XM_023651538.1 | XP_023507306.1 | 1022 | 79 | |||
| XM_023651539.1 | XP_023507307.1 | 508 | 79 | |||
Set Membership: |
|