Search
Search

SLC17A5| Gene Symbol: SLC17A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498010.4 | XP_001498060.3 | 1503 | 71 | ||
| XM_023650656.1 | XP_023506424.1 | 1463 | 71 | |||
| XM_023650657.1 | XP_023506425.1 | 1435 | 71 | |||
Set Membership: |
|