Search
Search

MAGI3| Gene Symbol: MAGI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014740072.2 | XP_014595558.1 | 1079 | 86 | ||
| XM_023642107.1 | XP_023497875.1 | 1209 | 86 | |||
| XM_023642108.1 | XP_023497876.1 | 1204 | 86 | |||
| XM_023642109.1 | XP_023497877.1 | 1073 | 86 | |||
| XM_023642110.1 | XP_023497878.1 | 1211 | 86 | |||
| XM_023642111.1 | XP_023497879.1 | 608 | 86 | |||
| XM_023642112.1 | XP_023497880.1 | 1210 | 86 | |||
| XM_023642114.1 | XP_023497882.1 | 1079 | 86 | |||
| XM_023642115.1 | XP_023497883.1 | 1073 | 86 | |||
| GenBank mRNA | JL632424.1 | - | 1 - 2 | 103 | 86 | |
Set Membership: |
|