Search
Search

AMPD2| Gene Symbol: AMPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023641458.1 | XP_023497226.1 | 2500 | 72 | ||
| XM_023641459.1 | XP_023497227.1 | 2444 | 72 | |||
| XM_023641460.1 | XP_023497228.1 | 2371 | 72 | |||
| XM_023641461.1 | XP_023497229.1 | 2017 | 72 | |||
| XM_023641462.1 | XP_023497230.1 | 2315 | 72 | |||
| XM_023641463.1 | XP_023497231.1 | 2502 | 72 | |||
Set Membership: |
|