Search
Search

PSRC1| Gene Symbol: PSRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023641483.1 | XP_023497251.1 | 338 | 62 | ||
| XM_023641484.1 | XP_023497252.1 | 337 | 62 | |||
| XM_023641486.1 | XP_023497254.1 | 333 | 62 | |||
| XM_023641487.1 | XP_023497255.1 | 334 | 62 | |||
| XM_023641488.1 | XP_023497256.1 | 226 | 62 | |||
Set Membership: |
|