Search
Search

ENDOV| Gene Symbol: ENDOV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023651849.1 | XP_023507617.1 | 592 | 86 | ||
| XM_023651850.1 | XP_023507618.1 | 591 | 86 | |||
| XM_023651851.1 | XP_023507619.1 | 593 | 86 | |||
| XM_023651852.1 | XP_023507620.1 | 459 | 86 | |||