Search
Search

ABCA10| Gene Symbol: ABCA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493969.6 | XP_001494019.3 | 4295 | 68 | ||
| XM_014739132.2 | XP_014594618.1 | 4241 | 68 | |||
| XM_014739136.2 | XP_014594622.1 | 4295 | 68 | |||
| XM_023652143.1 | XP_023507911.1 | 4340 | 68 | |||
| XM_023652144.1 | XP_023507912.1 | 4124 | 68 | |||
Set Membership: |
|