Search
Search

RPH3AL| Gene Symbol: RPH3AL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005597647.3 | XP_005597704.2 | 734 | 83 | ||
| XM_014727779.2 | XP_014583265.1 | 746 | 83 | |||
| XM_014727785.2 | XP_014583271.1 | 746 | 83 | |||
| XM_014727789.2 | XP_014583275.1 | 659 | 83 | |||
| XM_014727792.2 | XP_014583278.1 | 603 | 83 | |||
| XM_023653193.1 | XP_023508961.1 | 743 | 83 | |||
| XM_023653194.1 | XP_023508962.1 | 735 | 83 | |||
| XM_023653195.1 | XP_023508963.1 | 778 | 83 | |||
Set Membership: |
|