Search
Search

P2RX1| Gene Symbol: P2RX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014728014.2 | XP_014583500.1 | 863 | 76 | ||
| XM_014728020.2 | XP_014583506.1 | 865 | 76 | |||
| XM_014728022.2 | XP_014583508.1 | 867 | 76 | |||
| GenBank mRNA | KC790392.1 | - | 2 - 3 | 224 | 76 | |
Set Membership: |
|