Search
Search

CHRNB1| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503088.5 | XP_001503138.2 | 1315 | 94 | ||
| XM_005597750.3 | XP_005597807.1 | 1288 | 94 | |||
| XM_005597751.3 | XP_005597808.1 | 1270 | 94 | |||
| XM_005597752.3 | XP_005597809.1 | 1204 | 94 | |||
| XM_014728557.2 | XP_014584043.1 | 1280 | 94 | |||
Set Membership: |
|