Search
Search

BEST1| Gene Symbol: BEST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502370.4 | XP_001502420.2 | 1186 | 123 | ||
| XM_014729609.2 | XP_014585095.1 | 1541 | 123 | |||
| XM_023654196.1 | XP_023509964.1 | 998 | 123 | |||
Set Membership: |
|