Search
Search

DDAH1| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005610458.3 | XP_005610515.2 | 693 | 69 | ||
| XM_023641740.1 | XP_023497508.1 | 398 | 69 | |||
| XM_023641741.1 | XP_023497509.1 | 386 | 69 | |||
Set Membership: |
|