Search
Search

SGIP1| Gene Symbol: SGIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014740204.2 | XP_014595690.2 | 1985 | 67 | ||
| XM_023641917.1 | XP_023497685.1 | 1845 | 67 | |||
| XM_023641918.1 | XP_023497686.1 | 1833 | 67 | |||
| XM_023641919.1 | XP_023497687.1 | 3641 | 67 | |||
| XM_023641920.1 | XP_023497688.1 | 1785 | 67 | |||
| XM_023641921.1 | XP_023497689.1 | 1773 | 67 | |||
| XM_023641922.1 | XP_023497690.1 | 2231 | 67 | |||
| XM_023641923.1 | XP_023497691.1 | 1942 | 67 | |||
| XM_023641924.1 | XP_023497692.1 | 1948 | 67 | |||
| XM_023641925.1 | XP_023497693.1 | 1934 | 67 | |||
| XM_023641926.1 | XP_023497694.1 | 1913 | 67 | |||
| XM_023641927.1 | XP_023497695.1 | 1281 | 67 | |||
| XM_023641928.1 | XP_023497696.1 | 1257 | 67 | |||
| XM_023641929.1 | XP_023497697.1 | 1197 | 67 | |||
Set Membership: |
|