Search
Search

SPAG16| Gene Symbol: SPAG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014740282.2 | XP_014595768.2 | 612 | 96 | ||
| XM_023642274.1 | XP_023498042.1 | 614 | 96 | |||
| XM_023642277.1 | XP_023498045.1 | 1039 | 96 | |||
Set Membership: |
|