Search
Search

SPTBN2| Gene Symbol: SPTBN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654509.1 | XP_023510277.1 | 2962 | 58 | ||
| XM_023654510.1 | XP_023510278.1 | 3178 | 58 | |||
| XM_023654512.1 | XP_023510280.1 | 3117 | 58 | |||
| XM_023654513.1 | XP_023510281.1 | 2967 | 58 | |||
Set Membership: |
|