Search
Search

SYT12| Gene Symbol: SYT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001497062.6 | XP_001497112.1 | 915 | 71 | ||
| XM_023654536.1 | XP_023510304.1 | 909 | 71 | |||
| XM_023654537.1 | XP_023510305.1 | 959 | 71 | |||
| XM_023654539.1 | XP_023510307.1 | 866 | 71 | |||
Set Membership: |
|