Search
Search

KMT5B| Gene Symbol: KMT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654594.1 | XP_023510362.1 | 1181 | 85 | ||
| XM_023654595.1 | XP_023510363.1 | 1063 | 85 | |||
| XM_023654596.1 | XP_023510364.1 | 1073 | 85 | |||
| XM_023654597.1 | XP_023510365.1 | 1112 | 85 | |||
| XM_023654598.1 | XP_023510366.1 | 696 | 85 | |||
| XM_023654599.1 | XP_023510367.1 | 616 | 85 | |||
| XM_023654600.1 | XP_023510368.1 | 1181 | 85 | |||