Search
Search

OSBPL5| Gene Symbol: OSBPL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023654725.1 | XP_023510493.1 | 443 | 63 | ||
| XM_023654727.1 | XP_023510495.1 | 458 | 63 | |||
| XM_023654728.1 | XP_023510496.1 | 524 | 63 | |||
Set Membership: |
|