Search
Search

C13H7orf26| Gene Symbol: C13H7orf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655231.1 | XP_023510999.1 | 580 | 55 | ||
| XM_023655232.1 | XP_023511000.1 | 727 | 55 | |||
| XM_023655233.1 | XP_023511001.1 | 766 | 55 | |||
Set Membership: |
|