Search
Search

RNF216| Gene Symbol: RNF216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655205.1 | XP_023510973.1 | 1930 | 61 | ||
| XM_023655206.1 | XP_023510974.1 | 1759 | 61 | |||
| XM_023655207.1 | XP_023510975.1 | 1930 | 61 | |||
Set Membership: |
|