Search
Search

BRAT1| Gene Symbol: BRAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655160.1 | XP_023510928.1 | 2423 | 64 | ||
| XM_023655161.1 | XP_023510929.1 | 2508 | 64 | |||
| XM_023655162.1 | XP_023510930.1 | 2371 | 64 | |||
| XM_023655163.1 | XP_023510931.1 | 2091 | 64 | |||
| XM_023655164.1 | XP_023510932.1 | 2498 | 64 | |||
| XM_023655165.1 | XP_023510933.1 | 2379 | 64 | |||
| XM_023655166.1 | XP_023510934.1 | 2588 | 64 | |||
| XM_023655168.1 | XP_023510936.1 | 2353 | 64 | |||
| XM_023655169.1 | XP_023510937.1 | 2436 | 64 | |||
| XM_023655170.1 | XP_023510938.1 | 2343 | 64 | |||
| XM_023655171.1 | XP_023510939.1 | 2224 | 64 | |||
Set Membership: |
|