Search
Search

ZFAND2A| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003362685.4 | XP_003362733.2 | 520 | 87 | ||
| XM_005598554.3 | XP_005598611.1 | 528 | 87 | |||
| XM_023655091.1 | XP_023510859.1 | 528 | 87 | |||
| XM_023655093.1 | XP_023510861.1 | 517 | 87 | |||
Set Membership: |
|