Search
Search
| Gene Symbol: SUN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655066.1 | XP_023510834.1 | 801 | 77 | ||
| XM_023655068.1 | XP_023510836.1 | 801 | 77 | |||
| XM_023655069.1 | XP_023510837.1 | 801 | 77 | |||
Set Membership: |
|