Search
Search

GTF2IRD1| Gene Symbol: GTF2IRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023655487.1 | XP_023511255.1 | 1615 | 112 | ||
| XM_023655488.1 | XP_023511256.1 | 2237 | 112 | |||
| XM_023655489.1 | XP_023511257.1 | 1764 | 112 | |||
| XM_023655490.1 | XP_023511258.1 | 2088 | 112 | |||
| XM_023655491.1 | XP_023511259.1 | 1615 | 112 | |||
| XM_023655492.1 | XP_023511260.1 | 1447 | 112 | |||
| XM_023655493.1 | XP_023511261.1 | 1904 | 112 | |||
| XM_023655494.1 | XP_023511262.1 | 1652 | 112 | |||
| XM_023655495.1 | XP_023511263.1 | 1447 | 112 | |||
Set Membership: |
|