Search
Search

SUMF2| Gene Symbol: SUMF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001493334.6 | XP_001493384.5 | 160 | 68 | ||
| XM_014729945.2 | XP_014585431.2 | 160 | 68 | |||
| XM_023655547.1 | XP_023511315.1 | 160 | 68 | |||
| XM_023655549.1 | XP_023511317.1 | 160 | 68 | |||
Set Membership: |
|